Tisk…

Anti-EGF Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9695-100EA 28960 CZK
ABCAAB9695-100
Anti-EGF Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EGF (epidermal growth factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR