Tisk…

Anti-MELK Goat Polyclonal Antibody

ABCAAB9696-100EA 16480 CZK
ABCAAB9696-100
Anti-MELK Goat Polyclonal Antibody
Type: Primary
Antigen: MELK (maternal embryonic leucine zipper kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR