Tisk…
Anti-AKAP12 Goat Polyclonal Antibody
Dodavatel: Abcam
ABCAAB9698-100EA 21330 CZK
ABCAAB9698-100
Anti-AKAP12 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AKAP12 (A kinase (PRKA) anchor protein 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human