Tisk…

Anti-CCL5 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9783-100EA 21850 CZK
ABCAAB9783-100
Anti-CCL5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CCL5 (chemokine (C-C motif) ligand 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR