Tisk…

Anti-CNTF Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9785-100EA 22860 CZK
ABCAAB9785-100
Anti-CNTF Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CNTF (ciliary neurotrophic factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR