Tisk…

Anti-CXCL5 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9802-100EA 27640 CZK
ABCAAB9802-100
Anti-CXCL5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CXCL5 (chemokine (C-X-C motif) ligand 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR