Tisk…

Anti-ADAM11 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9847-100EA 26970 CZK
ABCAAB9847-100
Anti-ADAM11 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ADAM11 (ADAM metallopeptidase domain 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR