Tisk…

Anti-CCL1 Rabbit Polyclonal Antibody

ABCAAB9853-100EA 15370 CZK
ABCAAB9853-100
Anti-CCL1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: CCL1 (chemokine (C-C motif) ligand 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR