Tisk…

Anti-ADAM11 Mouse Monoclonal Antibody

Dodavatel: Abcam
ABCAAB9857-500EA 21140 CZK
ABCAAB9857-500
Anti-ADAM11 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ADAM11 (ADAM metallopeptidase domain 11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR