Tisk…

Anti-SOCS1 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9870-100EA 23810 CZK
ABCAAB9870-100
Anti-SOCS1 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SOCS1 (suppressor of cytokine signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR