Tisk…

Anti-IL1A Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9875-100EA 22310 CZK
ABCAAB9875-100
Anti-IL1A Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: IL1A (interleukin 1, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR