Tisk…

Anti-Interferon gamma Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB9918-100EA 21140 CZK
ABCAAB9918-100
Anti-Interferon gamma Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Interferon gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR