Tisk…

Anti-GFAP Mouse Monoclonal Antibody [clone: GF5]

Dodavatel: Abcam
ABCAAB10062-100EA 22830 CZK
ABCAAB10062-100
Anti-GFAP Mouse Monoclonal Antibody [clone: GF5]
Protilátky
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: monoclonal
Clone: GF5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR