Tisk…

Anti-MUC1 Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: M3A106]

ABCAAB10119-200EA 16710 CZK
ABCAAB10119-200
Anti-MUC1 Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: M3A106]
Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: monoclonal
Clone: M3A106
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR