Tisk…

Anti-PKA R2 Mouse Monoclonal Antibody [clone: RS6]

ABCAAB10128-500EA 17160 CZK
ABCAAB10128-500
Anti-PKA R2 Mouse Monoclonal Antibody [clone: RS6]
Type: Primary
Antigen: PKA R2
Clonality: monoclonal
Clone: RS6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR