Tisk…

Anti-ACVRL1 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB10179-100EA 22520 CZK
ABCAAB10179-100
Anti-ACVRL1 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ACVRL1 (activin A receptor type II-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR