Tisk…
Anti-Prostate Specific Antigen Mouse Monoclonal Antibody [clone: 8A6]
Dodavatel: Abcam
ABCAAB10187-250EA 21140 CZK
ABCAAB10187-250
Anti-Prostate Specific Antigen Mouse Monoclonal Antibody [clone: 8A6]
Protilátky
Type: Primary
Antigen: Prostate Specific Antigen
Clonality: monoclonal
Clone: 8A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human