Tisk…

Anti-PAPPA Mouse Monoclonal Antibody [clone: 10A5]

ABCAAB10193-250EA 17160 CZK
ABCAAB10193-250
Anti-PAPPA Mouse Monoclonal Antibody [clone: 10A5]
Type: Primary
Antigen: PAPPA (pregnancy-associated plasma protein A, pappalysin 1)
Clonality: monoclonal
Clone: 10A5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR