Tisk…

Anti-PAPPA Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: 10H9]

ABCAAB10195-200EA 17330 CZK
ABCAAB10195-200
Anti-PAPPA Mouse Monoclonal Antibody HRP (Horseradish Peroxidase) [clone: 10H9]
Type: Primary
Antigen: PAPPA (pregnancy-associated plasma protein A, pappalysin 1)
Clonality: monoclonal
Clone: 10H9
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR