Tisk…
Anti-TNNI3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: MF4]
Dodavatel: Abcam
ABCAAB10239-200EA 25560 CZK
ABCAAB10239-200
Anti-TNNI3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: MF4]
Protilátky
Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: monoclonal
Clone: MF4
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat