Tisk…

Anti-FITC Mouse Monoclonal Antibody [clone: 2A3]

Dodavatel: Abcam
ABCAAB10257-500EA 22340 CZK
ABCAAB10257-500
Anti-FITC Mouse Monoclonal Antibody [clone: 2A3]
Protilátky
Type: Primary
Antigen: FITC
Clonality: monoclonal
Clone: 2A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR