Tisk…

Anti-CCL8 Goat Polyclonal Antibody

ABCAAB10392-25EA 16080 CZK
ABCAAB10392-25
Anti-CCL8 Goat Polyclonal Antibody
Type: Primary
Antigen: CCL8 (chemokine (C-C motif) ligand 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR