Tisk…

Anti-CCL5 Goat Polyclonal Antibody

ABCAAB10393-100EA 16280 CZK
ABCAAB10393-100
Anti-CCL5 Goat Polyclonal Antibody
Type: Primary
Antigen: CCL5 (chemokine (C-C motif) ligand 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR