Tisk…

Anti-SMC2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB10411-50EA 21540 CZK
ABCAAB10411-50
Anti-SMC2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SMC2 (structural maintenance of chromosomes 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR