Tisk…

Anti-EGFR Mouse Monoclonal Antibody [clone: 29.1]

ABCAAB10414-100EA 16280 CZK
ABCAAB10414-100
Anti-EGFR Mouse Monoclonal Antibody [clone: 29.1]
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone: 29.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR