Tisk…
Anti-BNIP3 Mouse Monoclonal Antibody [clone: ANa40]
Dodavatel: Abcam
ABCAAB10433-100EA 28680 CZK
ABCAAB10433-100
Anti-BNIP3 Mouse Monoclonal Antibody [clone: ANa40]
Protilátky
Type: Primary
Antigen: BNIP3 (BCL2/adenovirus E1B 19kDa interacting protein 3)
Clonality: monoclonal
Clone: ANa40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat