Tisk…

Anti-CASP7 Rabbit Polyclonal Antibody

ABCAAB10449-100EA 16280 CZK
ABCAAB10449-100
Anti-CASP7 Rabbit Polyclonal Antibody
Type: Primary
Antigen: CASP7 (caspase 7, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR