Tisk…
Anti-CKII alpha prime polypeptide Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB10474-50EA 28500 CZK
ABCAAB10474-50
Anti-CKII alpha prime polypeptide Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CKII alpha prime polypeptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human