Tisk…
Anti-CBX1 / HP1 beta Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB10478-100EA 31560 CZK
ABCAAB10478-100
Anti-CBX1 / HP1 beta Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CBX1 / HP1 beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat