Tisk…

Anti-HPRT1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB10479-100EA 25860 CZK
ABCAAB10479-100
Anti-HPRT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HPRT1 (hypoxanthine phosphoribosyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR