Tisk…
Anti-TCERG1 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB10481-50EA 26870 CZK
ABCAAB10481-50
Anti-TCERG1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TCERG1 (transcription elongation regulator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human