Tisk…
Anti-LGALS7 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB10482-50EA 28500 CZK
ABCAAB10482-50
Anti-LGALS7 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LGALS7 (lectin, galactoside-binding, soluble, 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse