Tisk…

Anti-TRIM28 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB10483-50EA 23990 CZK
ABCAAB10483-50
Anti-TRIM28 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRIM28 (tripartite motif containing 28)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR