Tisk…
Anti-EP300 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB10485-100EA 28740 CZK
ABCAAB10485-100
Anti-EP300 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EP300 (E1A binding protein p300)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human