Tisk…

Anti-ROBO4 Rabbit Polyclonal Antibody

ABCAAB10547-100EA 16960 CZK
ABCAAB10547-100
Anti-ROBO4 Rabbit Polyclonal Antibody
Type: Primary
Antigen: ROBO4 (roundabout, axon guidance receptor, homolog 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR