Tisk…

Anti-GGA1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB10551-100EA 22860 CZK
ABCAAB10551-100
Anti-GGA1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GGA1 (golgi-associated, gamma adaptin ear containing, ARF binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR