Tisk…

Anti-CCL7 Mouse Monoclonal Antibody [clone: 36320.11]

ABCAAB10589-100EA 15650 CZK
ABCAAB10589-100
Anti-CCL7 Mouse Monoclonal Antibody [clone: 36320.11]
Type: Primary
Antigen: CCL7 (chemokine (C-C motif) ligand 7)
Clonality: monoclonal
Clone: 36320.11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR