Tisk…

Anti-Interferon beta Goat Polyclonal Antibody

ABCAAB10740-50EA 16280 CZK
ABCAAB10740-50
Anti-Interferon beta Goat Polyclonal Antibody
Type: Primary
Antigen: Interferon beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR