Tisk…

Anti-CCN1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB10760-100EA 21850 CZK
ABCAAB10760-100
Anti-CCN1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CCN1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR