Tisk…

Anti-CAPN9 Rabbit Polyclonal Antibody

ABCAAB10824-50EA 15650 CZK
ABCAAB10824-50
Anti-CAPN9 Rabbit Polyclonal Antibody
Type: Primary
Antigen: CAPN9 (calpain 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR