Tisk…

Anti-PDGFRB Goat Polyclonal Antibody

ABCAAB10848-25EA 17250 CZK
ABCAAB10848-25
Anti-PDGFRB Goat Polyclonal Antibody
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR