Tisk…

Anti-Cdc13 Mouse Monoclonal Antibody [clone: 6F11/2]

Dodavatel: Abcam
ABCAAB10873-50EA 22340 CZK
ABCAAB10873-50
Anti-Cdc13 Mouse Monoclonal Antibody [clone: 6F11/2]
Protilátky
Type: Primary
Antigen: Cdc13
Clonality: monoclonal
Clone: 6F11/2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR