Tisk…

Anti-RB1 Rat Monoclonal Antibody [clone: 7F10]

ABCAAB10922-50EA 16080 CZK
ABCAAB10922-50
Anti-RB1 Rat Monoclonal Antibody [clone: 7F10]
Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: monoclonal
Clone: 7F10
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR