Tisk…

Anti-MYB Mouse Monoclonal Antibody [clone: IOA3]

ABCAAB10937-50EA 17250 CZK
ABCAAB10937-50
Anti-MYB Mouse Monoclonal Antibody [clone: IOA3]
Type: Primary
Antigen: MYB (v-myb avian myeloblastosis viral oncogene homolog)
Clonality: monoclonal
Clone: IOA3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR