Tisk…

Anti-ATR Rabbit Polyclonal Antibody

ABCAAB10946-50EA 15650 CZK
ABCAAB10946-50
Anti-ATR Rabbit Polyclonal Antibody
Type: Primary
Antigen: ATR (ataxia telangiectasia and Rad3 related)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR