Tisk…

Anti-SLIT1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB10984-100EA 22520 CZK
ABCAAB10984-100
Anti-SLIT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SLIT1 (slit homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR