Tisk…

Anti-AQP1 Mouse Monoclonal Antibody [clone: 1/A5F6]

Dodavatel: Abcam
ABCAAB11023-50EA 21330 CZK
ABCAAB11023-50
Anti-AQP1 Mouse Monoclonal Antibody [clone: 1/A5F6]
Protilátky
Type: Primary
Antigen: AQP1 (aquaporin 1 (Colton blood group))
Clonality: monoclonal
Clone: 1/A5F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR