Tisk…

Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9]

Dodavatel: Abcam
ABCAAB11029-100EA 23660 CZK
ABCAAB11029-100
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9]
Protilátky
Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: monoclonal
Clone: 3.9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR