Tisk…

Anti-CUL1 Mouse Monoclonal Antibody [clone: AS97.1]

Dodavatel: Abcam
ABCAAB11047-50EA 26870 CZK
ABCAAB11047-50
Anti-CUL1 Mouse Monoclonal Antibody [clone: AS97.1]
Protilátky
Type: Primary
Antigen: CUL1 (cullin 1)
Clonality: monoclonal
Clone: AS97.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR