Tisk…

Anti-Histone H1.0 Mouse Monoclonal Antibody [clone: 27]

Dodavatel: Abcam
ABCAAB11080-100EA 23320 CZK
ABCAAB11080-100
Anti-Histone H1.0 Mouse Monoclonal Antibody [clone: 27]
Protilátky
Type: Primary
Antigen: Histone H1.0
Clonality: monoclonal
Clone: 27
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR